Assay Details
Gene Symbol: slc25a3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328572.1 | NP_001315501.1 | 4 - 5 | 534 | 70 | |
NM_200715.2 | NP_957009.1 | 4 - 5 | 498 | 70 | ||
XM_005164728.4 | XP_005164785.1 | 4 - 5 | 700 | 70 | ||
GenBank mRNA | BC059476.1 | - | 4 - 5 | 475 | 70 | |
EH451459.1 | - | 4 - 5 | 524 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
