Assay Details
Gene Symbol: slc25a3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328572.1 | NP_001315501.1 | 5 - 6 | 711 | 77 | |
NM_200715.2 | NP_957009.1 | 5 - 6 | 675 | 77 | ||
XM_005164728.4 | XP_005164785.1 | 5 - 6 | 877 | 77 | ||
GenBank mRNA | BC059476.1 | - | 5 - 6 | 77 | ||
EH451459.1 | - | 5 - 6 | 704 | 78 | ||
EH478503.1 | - | 3 - 4 | 712 | 79 | ||
EH609753.1 | - | 3 - 4 | 710 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
