Assay Details
| Gene Symbol: xrcc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200786.1 | NP_957080.1 | 3 - 4 | 435 | 67 | |
| XM_009305682.2 | XP_009303957.1 | 3 - 4 | 460 | 67 | ||
| GenBank mRNA | BC059635.1 | - | 3 - 4 | 435 | 67 | |
More Information
Set Membership: |
|

xrcc4
Genomic Map