Assay Details
| Gene Symbol: gnao1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200828.1 | NP_957122.1 | 1 - 2 | 200 | 70 | |
| GenBank mRNA | BC060909.1 | - | 1 - 2 | 200 | 70 | |
More Information
Set Membership: |
|

gnao1b
Genomic Map