Assay Details
| Gene Symbol: gnao1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200828.1 | NP_957122.1 | 5 - 6 | 675 | 95 | |
| GenBank mRNA | BC060909.1 | - | 5 - 6 | 675 | 95 | |
More Information
Set Membership: |
|

gnao1b
Genomic Map