Assay Details
| Gene Symbol: eif3ea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200839.1 | NP_957133.1 | 12 - 13 | 1322 | 71 | |
| GenBank mRNA | AY398340.1 | - | 12 - 13 | 1313 | 71 | |
| AY648778.1 | - | 12 - 13 | 1321 | 71 | ||
| BC061454.1 | - | 12 - 13 | 1322 | 71 | ||
More Information
Set Membership: |
|

eif3ea
Genomic Map