Assay Details
| Gene Symbol: abhd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200914.1 | NP_957208.1 | 4 - 5 | 653 | 58 | |
| GenBank mRNA | BC047183.1 | - | 4 - 5 | 653 | 58 | |
| BC165808.1 | - | 2 - 3 | 421 | 58 | ||
More Information
Set Membership: |
|

abhd2a
Genomic Map