Assay Details
Gene Symbol: prkcba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201029.1 | NP_957323.1 | 2 - 3 | 312 | 61 | |
XM_005170844.4 | XP_005170901.1 | 2 - 3 | 357 | 61 | ||
GenBank mRNA | BC054603.1 | - | 2 - 3 | 312 | 61 | |
BC171680.1 | - | 2 - 3 | 276 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
