Assay Details
Gene Symbol: rps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201146.1 | NP_957440.1 | 3 - 4 | 354 | 144 | |
GenBank mRNA | BC055098.1 | - | 3 - 4 | 354 | 144 | |
BC067658.1 | - | 2 - 3 | 343 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
