Assay Details
| Gene Symbol: rps10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201146.1 | NP_957440.1 | 4 - 5 | 429 | 74 | |
| GenBank mRNA | BC055098.1 | - | 4 - 5 | 429 | 74 | |
| BC067658.1 | - | 3 - 4 | 418 | 74 | ||
More Information
Set Membership: |
|

rps10
Genomic Map