Assay Details
| Gene Symbol: rybpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201317.2 | NP_958474.2 | 3 - 4 | 898 | 106 | |
| GenBank mRNA | BC054937.1 | - | 3 - 4 | 901 | 106 | |
More Information
Set Membership: |
|

rybpa
Genomic Map