Assay Details
| Gene Symbol: rdh10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201331.1 | NP_958488.1 | 1 - 2 | 397 | 82 | |
| GenBank mRNA | BC054596.1 | - | 1 - 2 | 397 | 82 | |
| BC164370.1 | - | 1 - 2 | 307 | 82 | ||
More Information
Set Membership: |
|

rdh10b
Genomic Map