Assay Details
| Gene Symbol: rdh10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201331.1 | NP_958488.1 | 3 - 4 | 732 | 98 | |
| GenBank mRNA | AM422096.1 | - | 2 - 3 | 301 | 98 | |
| BC054596.1 | - | 3 - 4 | 732 | 98 | ||
| BC164370.1 | - | 3 - 4 | 642 | 98 | ||
More Information
Set Membership: |
|

rdh10b
Genomic Map