Assay Details
| Gene Symbol: ldb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201505.1 | NP_958913.1 | 6 - 7 | 690 | 111 | |
| GenBank mRNA | BC045908.1 | - | 6 - 7 | 690 | 111 | |
| BC164050.1 | - | 5 - 6 | 588 | 111 | ||
| DQ012165.1 | - | 5 - 6 | 554 | 111 | ||
More Information
Set Membership: |
|

ldb3a
Genomic Map