Assay Details
| Gene Symbol: ldb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201505.1 | NP_958913.1 | 8 - 9 | 877 | 64 | |
| XM_021480673.1 | XP_021336348.1 | 7 - 8 | 954 | 64 | ||
| GenBank mRNA | BC045908.1 | - | 8 - 9 | 877 | 64 | |
| BC164050.1 | - | 7 - 8 | 775 | 64 | ||
| DQ012162.1 | - | 5 - 6 | 765 | 64 | ||
| DQ012165.1 | - | 7 - 8 | 741 | 64 | ||
| DQ012166.1 | - | 6 - 7 | 717 | 64 | ||
More Information
Set Membership: |
|

ldb3a
Genomic Map