Assay Details
| Gene Symbol: ldb3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201505.1 | NP_958913.1 | 9 - 10 | 914 | 65 | |
| XM_021480673.1 | XP_021336348.1 | 8 - 9 | 991 | 65 | ||
| GenBank mRNA | BC045908.1 | - | 9 - 10 | 914 | 65 | |
| BC164050.1 | - | 8 - 9 | 812 | 65 | ||
| DQ012162.1 | - | 6 - 7 | 802 | 65 | ||
| DQ012165.1 | - | 8 - 9 | 778 | 65 | ||
| DQ012166.1 | - | 7 - 8 | 754 | 65 | ||
More Information
Set Membership: |
|

ldb3a
Genomic Map