Assay Details
Gene Symbol: nup50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201580.2 | NP_963874.2 | 5 - 6 | 988 | 63 | |
GenBank mRNA | AY398390.1 | - | 5 - 6 | 987 | 63 | |
BC051604.1 | - | 5 - 6 | 936 | 63 | ||
BC065637.1 | - | 5 - 6 | 988 | 63 | ||
BC153634.1 | - | 3 - 4 | 660 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
