Assay Details
| Gene Symbol: naa15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203321.2 | NP_976066.2 | 13 - 14 | 1699 | 89 | |
| GenBank mRNA | AY391464.1 | - | 13 - 14 | 1686 | 89 | |
| BC044392.1 | - | 13 - 14 | 1616 | 89 | ||
| BC071438.1 | - | 13 - 14 | 1699 | 89 | ||
More Information
Set Membership: |
|

naa15b
Genomic Map