Assay Details
| Gene Symbol: traf4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205762.1 | NP_991325.1 | 6 - 7 | 1034 | 53 | |
| GenBank mRNA | AJ556548.1 | - | 6 - 7 | 1097 | 53 | |
| BC065969.1 | - | 6 - 7 | 1034 | 53 | ||
More Information
Set Membership: |
|

traf4a
Genomic Map