Assay Details
| Gene Symbol: cmpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213109.2 | NP_998274.2 | 2 - 3 | 445 | 147 | |
| GenBank mRNA | BC049446.1 | - | 2 - 3 | 375 | 147 | |
| EH555529.1 | - | 2 - 3 | 484 | 147 | ||
More Information
Set Membership: |
|

cmpk
Genomic Map