Assay Details
| Gene Symbol: cmpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213109.2 | NP_998274.2 | 5 - 6 | 768 | 68 | |
| GenBank mRNA | BC049446.1 | - | 5 - 6 | 698 | 68 | |
| EH461618.1 | - | 1 - 2 | 843 | 68 | ||
| EH555529.1 | - | 5 - 6 | 807 | 68 | ||
More Information
Set Membership: |
|

cmpk
Genomic Map