Assay Details
| Gene Symbol: yy1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213120.1 | NP_998285.1 | 3 - 4 | 820 | 80 | |
| XM_005170229.4 | XP_005170286.1 | 4 - 5 | 1091 | 80 | ||
| GenBank mRNA | BC053307.1 | - | 3 - 4 | 820 | 80 | |
More Information
Set Membership: |
|

yy1b
Genomic Map