Assay Details
| Gene Symbol: bcl7ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213165.1 | NP_998330.1 | 5 - 6 | 546 | 97 | |
| GenBank mRNA | BC067572.1 | - | 5 - 6 | 546 | 97 | |
More Information
Set Membership: |
|

bcl7ba
Genomic Map