Assay Details
| Gene Symbol: stk25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213308.1 | NP_998473.1 | 2 - 3 | 567 | 96 | |
| XM_005171203.4 | XP_005171260.1 | 2 - 3 | 610 | 96 | ||
| GenBank mRNA | BC045867.1 | - | 2 - 3 | 543 | 96 | |
| BC066512.1 | - | 2 - 3 | 567 | 96 | ||
| BC164728.1 | - | 2 - 3 | 300 | 96 | ||
More Information
Set Membership: |
|

stk25b
Genomic Map