Assay Details
| Gene Symbol: emc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213370.1 | NP_998535.1 | 3 - 4 | 538 | 125 | |
| GenBank mRNA | BC046080.1 | - | 3 - 4 | 538 | 125 | |
More Information
Set Membership: |
|

emc8
Genomic Map