Assay Details
| Gene Symbol: rap1ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243179.1 | NP_001230108.1 | 5 - 6 | 629 | 75 | |
| NM_213405.2 | NP_998570.2 | 6 - 7 | 727 | 75 | ||
| XM_021479589.1 | XP_021335264.1 | 4 - 5 | 2168 | 75 | ||
| XM_021479590.1 | XP_021335265.1 | 5 - 6 | 636 | 75 | ||
| GenBank mRNA | BC049510.1 | - | 6 - 7 | 695 | 75 | |
| EH543099.1 | - | 5 - 6 | 645 | 75 | ||
More Information
Set Membership: |
|

rap1ab
Genomic Map