Assay Details
Gene Symbol: slc25a3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213722.1 | NP_998887.1 | 2 - 3 | 227 | 142 | |
GenBank mRNA | BC046007.1 | - | 2 - 3 | 227 | 142 | |
BC067565.1 | - | 2 - 3 | 185 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
