Assay Details
| Gene Symbol: snrpd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002081.1 | NP_001002081.1 | 2 - 3 | 192 | 70 | |
| XM_021478237.1 | XP_021333912.1 | 2 - 3 | 267 | 70 | ||
| GenBank mRNA | BC071554.1 | - | 2 - 3 | 192 | 70 | |
More Information
Set Membership: |
|

snrpd3
Genomic Map