Assay Details
| Gene Symbol: rab1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002129.2 | NP_001002129.2 | 4 - 5 | 550 | 74 | |
| GenBank mRNA | BC071442.1 | - | 4 - 5 | 387 | 74 | |
| BC097013.1 | - | 4 - 5 | 493 | 74 | ||
| EH444733.1 | - | 4 - 5 | 499 | 74 | ||
More Information
Set Membership: |
|

rab1bb
Genomic Map