Assay Details
| Gene Symbol: narf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002342.1 | NP_001002342.1 | 8 - 9 | 1043 | 69 | |
| GenBank mRNA | BC075921.1 | - | 8 - 9 | 1043 | 69 | |
More Information
Set Membership: |
|

narf
Genomic Map