Assay Details
Gene Symbol: trappc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002742.2 | NP_001002742.1 | 4 - 5 | 380 | 61 | |
XM_005170575.4 | XP_005170632.1 | 4 - 5 | 437 | 61 | ||
XM_021469786.1 | XP_021325461.1 | 4 - 5 | 390 | 61 | ||
GenBank mRNA | BC076411.1 | - | 4 - 5 | 384 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
