Assay Details
| Gene Symbol: pdia8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003517.1 | NP_001003517.1 | 5 - 6 | 693 | 70 | |
| GenBank mRNA | BC077131.1 | - | 5 - 6 | 693 | 70 | |
More Information
Set Membership: |
|

pdia8
Genomic Map