Assay Details
| Gene Symbol: pip5k1bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004579.1 | NP_001004579.1 | 8 - 9 | 1142 | 122 | |
| XM_005166729.4 | XP_005166786.1 | 8 - 9 | 1225 | 122 | ||
| XM_009303781.3 | XP_009302056.1 | 8 - 9 | 1295 | 122 | ||
| GenBank mRNA | BC081646.1 | - | 8 - 9 | 1142 | 122 | |
More Information
Set Membership: |
|

pip5k1bb
Genomic Map