Assay Details
| Gene Symbol: gtf2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004596.1 | NP_001004596.1 | 2 - 3 | 297 | 63 | |
| GenBank mRNA | BC081602.1 | - | 2 - 3 | 297 | 63 | |
More Information
Set Membership: |
|

gtf2h1
Genomic Map