Assay Details
| Gene Symbol: spout1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004627.1 | NP_001004627.1 | 2 - 3 | 116 | 118 | |
| GenBank mRNA | BC081398.1 | - | 2 - 3 | 116 | 118 | |
More Information
Set Membership: |
|

spout1
Genomic Map