Assay Details
| Gene Symbol: cdhr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005402.1 | NP_001005402.1 | 4 - 5 | 347 | 154 | |
| XM_005156820.4 | XP_005156877.1 | 3 - 4 | 460 | 154 | ||
| XM_021480567.1 | XP_021336242.1 | 4 - 5 | 481 | 154 | ||
| GenBank mRNA | AY684930.1 | - | 4 - 5 | 347 | 154 | |
More Information
Set Membership: |
|

cdhr1a
Genomic Map