Assay Details
| Gene Symbol: cpt1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328192.1 | NP_001315121.1 | 2 - 3 | 467 | 66 | |
| GenBank mRNA | BC083470.1 | - | 2 - 3 | 324 | 66 | |
| EH509285.1 | - | 2 - 3 | 335 | 66 | ||
More Information
Set Membership: |
|

cpt1b
Genomic Map