Assay Details
| Gene Symbol: dennd6aa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005987.1 | NP_001005987.1 | 8 - 9 | 856 | 68 | |
| XM_021479600.1 | XP_021335275.1 | 8 - 9 | 868 | 68 | ||
| GenBank mRNA | BC083420.1 | - | 8 - 9 | 856 | 68 | |
More Information
Set Membership: |
|

dennd6aa
Genomic Map