Assay Details
Gene Symbol: rp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007353.1 | NP_001007354.1 | 1 - 2 | 242 | 90 | |
XM_009294316.3 | XP_009292591.1 | 1 - 2 | 297 | 90 | ||
GenBank mRNA | BC085521.1 | - | 1 - 2 | 242 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
