Assay Details
| Gene Symbol: xirp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012377.1 | NP_001012377.1 | 1 | 6950 | 133 | |
| XM_009298510.2 | XP_009296785.1 | 4 | 7157 | 133 | ||
| XM_021480063.1 | XP_021335738.1 | 8 | 7898 | 133 | ||
| GenBank mRNA | AY812746.1 | - | 1 | 6950 | 133 | |
More Information
Set Membership: |
|

xirp1
Genomic Map