Assay Details
| Gene Symbol: dlg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012388.1 | NP_001012388.1 | 15 - 16 | 1851 | 86 | |
| XM_005163675.4 | XP_005163732.1 | 17 - 18 | 86 | |||
| XM_005163677.4 | XP_005163734.1 | 16 - 17 | 86 | |||
| XM_009296934.3 | XP_009295209.1 | 19 - 20 | 86 | |||
| XM_009296951.3 | XP_009295226.1 | 19 - 20 | 86 | |||
| XM_009296993.3 | XP_009295268.1 | 14 - 15 | 86 | |||
| GenBank mRNA | AY819035.1 | - | 15 - 16 | 1851 | 86 | |
More Information
Set Membership: |
|

dlg1l
Genomic Map