Assay Details
| Gene Symbol: dlg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012388.1 | NP_001012388.1 | 16 - 17 | 1877 | 120 | |
| XM_009296951.3 | XP_009295226.1 | 20 - 21 | 2877 | 120 | ||
| GenBank mRNA | AY819035.1 | - | 16 - 17 | 1877 | 120 | |
More Information
Set Membership: |
|

dlg1l
Genomic Map