Assay Details
| Gene Symbol: dlg1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012388.1 | NP_001012388.1 | 3 - 4 | 425 | 76 | |
| XM_005163667.4 | XP_005163724.1 | 6 - 7 | 1326 | 76 | ||
| GenBank mRNA | AY819035.1 | - | 3 - 4 | 425 | 76 | |
More Information
Set Membership: |
|

dlg1l
Genomic Map