Assay Details
| Gene Symbol: ccdc106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013305.2 | NP_001013323.2 | 2 - 3 | 267 | 89 | |
| NM_001312892.1 | NP_001299821.1 | 3 - 4 | 224 | 89 | ||
| XM_021468054.1 | XP_021323729.1 | 3 - 4 | 421 | 89 | ||
| GenBank mRNA | BC090716.1 | - | 2 - 3 | 265 | 89 | |
More Information
Set Membership: |
|

ccdc106b
Genomic Map