Assay Details
| Gene Symbol: ccdc106b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013305.2 | NP_001013323.2 | 3 - 4 | 479 | 67 | |
| NM_001312892.1 | NP_001299821.1 | 4 - 5 | 436 | 67 | ||
| XM_021468054.1 | XP_021323729.1 | 4 - 5 | 633 | 67 | ||
| GenBank mRNA | BC090716.1 | - | 3 - 4 | 477 | 67 | |
More Information
Set Membership: |
|

ccdc106b
Genomic Map