Assay Details
| Gene Symbol: eif4g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014289.2 | NP_001014311.1 | 12 - 13 | 1488 | 66 | |
| GenBank mRNA | AB096100.1 | - | 12 - 13 | 1554 | 66 | |
| BC059195.1 | - | 12 - 13 | 1487 | 66 | ||
| BC165852.1 | - | 5 - 6 | 683 | 66 | ||
More Information
Set Membership: |
|

eif4g2a
Genomic Map