Assay Details
| Gene Symbol: eif4g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014289.2 | NP_001014311.1 | 14 - 15 | 1732 | 77 | |
| GenBank mRNA | AB096100.1 | - | 14 - 15 | 1798 | 77 | |
| BC059195.1 | - | 14 - 15 | 1731 | 77 | ||
| BC165852.1 | - | 7 - 8 | 927 | 77 | ||
More Information
Set Membership: |
|

eif4g2a
Genomic Map