Assay Details
Gene Symbol: eif4g2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014289.2 | NP_001014311.1 | 6 - 7 | 703 | 95 | |
GenBank mRNA | AB096100.1 | - | 6 - 7 | 769 | 95 | |
BC059195.1 | - | 6 - 7 | 702 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
