Assay Details
| Gene Symbol: impdh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014369.2 | NP_001014391.1 | 5 - 6 | 569 | 80 | |
| XM_005164717.4 | XP_005164774.1 | 6 - 7 | 1075 | 80 | ||
| XM_005164718.4 | XP_005164775.1 | 5 - 6 | 997 | 80 | ||
| XM_017354952.2 | XP_017210441.1 | 6 - 7 | 665 | 80 | ||
| GenBank mRNA | BC091790.1 | - | 5 - 6 | 80 | ||
| EH434249.1 | - | 5 - 6 | 571 | 80 | ||
More Information
Set Membership: |
|

impdh1b
Genomic Map