Assay Details
Gene Symbol: ptges | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014828.2 | NP_001014828.1 | 2 - 3 | 300 | 109 | |
GenBank mRNA | AY724691.1 | - | 2 - 3 | 193 | 109 | |
BC095159.1 | - | 2 - 3 | 300 | 109 | ||
BC164946.1 | - | 2 - 3 | 227 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
